This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01185359_mH
See other EYA4 GE Assays ›
Gene Symbol:
Eya4
Gene Name:
eyes absent 4 homolog (Drosophila)
Gene Aliases:
B130023L16Rik
Chromosome Location:
Chr.10: 23102962 - 23350856 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01185359_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eya4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010167.4 NP_034297.2 13 - 14 1422 106
GenBank mRNA AK041625.1 - 13 - 14 1423 106
BC120898.1 - 13 - 14 1406 40061597 106
BC120899.1 - 13 - 14 1406 40061599 106
Y17115.1 - 14 - 15 1533 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001868.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eya4

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
multicellular organism development
sensory perception of sound
chromatin modification
middle ear morphogenesis
molecular_function
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000013483.mm.1 NM_010167.5 Refseq
Affymetrix Mouse Clariom S Array PSR1000013483.mm.1 TC1000001868.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0