This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01185455_m1
See other ZMPSTE24 GE Assays ›
Gene Symbol:
Zmpste24
Gene Name:
zinc metallopeptidase, STE24
Gene Aliases:
A530043O15Rik, D030046F19, FACE1, Face-1, MADB, STE24, Ste24p
Chromosome Location:
Chr.4: 121059237 - 121098249 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01185455_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zmpste24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172700.2 NP_766288.1 6 - 7 1013 112
GenBank mRNA AJ487544.1 - 6 - 7 979 112
AK083566.1 - 6 - 7 1013 112
AK170946.1 - 6 - 7 972 112
BC023740.1 - 6 - 7 956 112
BC138578.1 - 6 - 7 881 8861598 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003451.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zmpste24

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
nuclear envelope organization
prenylated protein catabolic process
CAAX-box protein processing
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400028025.mm.1 NM_172700.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028025.mm.1 NM_172700.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400028025.mm.1 TC0400003451.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline