This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187108_m1
See other ECE1 GE Assays ›
Gene Symbol:
Ece1
Gene Name:
endothelin converting enzyme 1
Gene Aliases:
AW322500, ECE-1, ECE-1a, ECE-1b, ECE-1d
Chromosome Location:
Chr.4: 137862216 - 137965229 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187108_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ece1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199307.2 NP_955011.1 10 - 11 1347 67
GenBank mRNA AK134088.1 - 10 - 11 1348 67
AK163790.1 - 7 - 8 1260 67
AK205173.1 - 2 - 3 251 67
BC038057.1 - 1 - 2 161 67
BC060648.1 - 10 - 11 1287 6854311 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001714.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ece1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
positive regulation of receptor recycling
proteolysis
apoptotic process
heart development
regulation of blood pressure
positive regulation of cardiac muscle hypertrophy
substance P catabolic process
bradykinin catabolic process
calcitonin catabolic process
protein processing
peptide hormone processing
positive regulation of cAMP biosynthetic process
endothelin maturation
regulation of vasodilation
hormone catabolic process
embryonic digit morphogenesis
ear development
positive regulation of G-protein coupled receptor protein signaling pathway
pharyngeal system development
regulation of ERK1 and ERK2 cascade
endopeptidase activity
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400013910.mm.1 NM_199307.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400013910.mm.1 NM_199307.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400013910.mm.1 TC0400001714.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-twzkw:80/100.66.77.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0