This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187247_m1
See other EIF2B3 GE Assays ›
Gene Symbol:
Eif2b3
Gene Name:
eukaryotic translation initiation factor 2B, subunit 3
Gene Aliases:
1190002P15Rik
Chromosome Location:
Chr.4: 117019399 - 117086852 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187247_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2b3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111277.1 NP_001104747.1 4 - 5 533 71
NM_175135.4 NP_780344.2 4 - 5 533 71
GenBank mRNA AK145856.1 - 4 - 5 510 71
AK172302.1 - 4 - 5 483 71
BC115657.1 - 2 - 3 207 40103205 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001274.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
regulation of translation
response to heat
response to glucose
oligodendrocyte development
negative regulation of translational initiation in response to stress
response to peptide hormone
positive regulation of GTPase activity
cellular response to stimulus
translation initiation factor activity
guanyl-nucleotide exchange factor activity
translation factor activity, RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400009551.mm.1 TC0400001274.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400009554.mm.1 NM_001111277.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009554.mm.1 NM_001111277.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009554.mm.1 NM_175135.5 Refseq
Affymetrix Mouse Clariom S Array PSR0400009554.mm.1 TC0400001274.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0