This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187260_g1
See other MUTYH GE Assays ›
Gene Symbol:
Mutyh
Gene Name:
mutY homolog (E. coli)
Gene Aliases:
5730495A01Rik, Mutyha, Mutyhalpha, Mutyhb, Mutyhbeta, Mutyhc, Myh
Chromosome Location:
Chr.4: 116807682 - 116819439 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187260_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mutyh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159581.1 NP_001153053.1 8 - 9 622 60
NM_133250.2 NP_573513.2 7 - 8 529 60
GenBank mRNA AB117937.1 - 5 - 6 417 60
AB117938.1 - 6 - 7 491 60
AK017730.1 - 7 - 8 517 60
AK077546.1 - 7 - 8 526 60
AK156790.1 - 8 - 9 612 60
AY007717.1 - 6 - 7 485 60
BC057942.1 - 7 - 8 510 5320744 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001266.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mutyh

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
metabolic process
purine-specific mismatch base pair DNA N-glycosylase activity
catalytic activity
uracil DNA N-glycosylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
MutSalpha complex binding
DNA-3-methylbase glycosylase activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400009478.mm.1 NM_001159581.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009478.mm.1 NM_001159581.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009478.mm.1 NM_133250.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009478.mm.1 NM_133250.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400009478.mm.1 TC0400001266.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0