This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187629_m1
See other EIF4ENIF1 GE Assays ›
Gene Symbol:
Eif4enif1
Gene Name:
eukaryotic translation initiation factor 4E nuclear import factor 1
Gene Aliases:
2610509L04Rik, A930019J01Rik, AA410001, AU021239, Clast4, D11Ertd166e
Chromosome Location:
Chr.11: 3201598 - 3244588 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187629_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4enif1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166547.1 NP_001160019.1 12 - 13 1989 83
NM_001166548.1 NP_001160020.1 11 - 12 1921 83
NM_001166549.1 NP_001160021.1 11 - 12 2025 83
NM_023743.2 NP_076232.2 12 - 13 2004 83
GenBank mRNA AB031388.1 - 12 - 13 1825 83
AK012082.1 - 2 - 3 408 83
AK041598.1 - 11 - 12 1883 83
AK088425.1 - 7 - 8 1091 83
AK128990.1 - 11 - 12 2023 83
AK165678.1 - 12 - 13 1991 83
AK209533.1 - 1 - 2 151 83
BC033410.1 - 12 - 13 1839 5359290 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000005.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif4enif1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
negative regulation of translation
stem cell population maintenance
negative regulation of neuron differentiation
mRNA binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000091.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000094.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000094.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000094.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000094.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000094.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000096.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000099.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000090.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000097.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000089.mm.1 TC1100000005.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0