This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187631_m1
See other EIF4ENIF1 GE Assays ›
Gene Symbol:
Eif4enif1
Gene Name:
eukaryotic translation initiation factor 4E nuclear import factor 1
Gene Aliases:
2610509L04Rik, A930019J01Rik, AA410001, AU021239, Clast4, D11Ertd166e
Chromosome Location:
Chr.11: 3201598 - 3244588 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187631_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4enif1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166547.1 NP_001160019.1 15 - 16 2301 69
NM_001166548.1 NP_001160020.1 14 - 15 2233 69
NM_001166549.1 NP_001160021.1 14 - 15 2337 69
NM_023743.2 NP_076232.2 15 - 16 2316 69
GenBank mRNA AK012082.1 - 5 - 6 720 69
AK041598.1 - 14 - 15 2195 69
AK088425.1 - 10 - 11 1403 69
AK128990.1 - 14 - 15 2335 69
AK165678.1 - 15 - 16 2303 69
BC033410.1 - 15 - 16 2151 5359290 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000005.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif4enif1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
negative regulation of translation
stem cell population maintenance
negative regulation of neuron differentiation
mRNA binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000104.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000105.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000106.mm.1 TC1100000005.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 NM_001166547.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 NM_001166547.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 NM_001166548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 NM_001166548.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 NM_001166549.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 NM_001166549.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 NM_023743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 NM_023743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100000103.mm.1 TC1100000005.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0