This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187752_g1
See other EWSR1 GE Assays ›
Gene Symbol:
Ewsr1
Gene Name:
Ewing sarcoma breakpoint region 1
Gene Aliases:
Ews, Ewsh
Chromosome Location:
Chr.11: 5069687 - 5099099 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187752_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ewsr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283061.1 NP_001269990.1 5 - 6 477 65
NM_001283062.1 NP_001269991.1 5 - 6 477 65
NM_001283063.1 NP_001269992.1 5 - 6 477 65
NM_007968.3 NP_031994.2 5 - 6 477 65
GenBank mRNA AK019460.1 - 5 - 6 426 65
AK034755.1 - 5 - 6 453 65
AK049743.1 - 5 - 6 457 65
AK146791.1 - 5 - 6 453 65
AK147103.1 - 5 - 6 444 65
AK147909.1 - 5 - 6 434 65
AK150345.1 - 5 - 6 435 65
AK151625.1 - 5 - 6 448 65
AK153276.1 - 5 - 6 435 65
BC068226.1 - 5 - 6 444 5707017 65
X79233.1 - 5 - 6 466 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ewsr1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
nucleic acid binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0