This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187753_g1
See other EWSR1 GE Assays ›
Gene Symbol:
Ewsr1
Gene Name:
Ewing sarcoma breakpoint region 1
Gene Aliases:
Ews, Ewsh
Chromosome Location:
Chr.11: 5069687 - 5099099 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187753_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ewsr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283061.1 NP_001269990.1 6 - 7 644 62
NM_001283062.1 NP_001269991.1 6 - 7 644 62
NM_001283063.1 NP_001269992.1 6 - 7 644 62
NM_007968.3 NP_031994.2 6 - 7 644 62
GenBank mRNA AK019460.1 - 6 - 7 593 62
AK034755.1 - 6 - 7 620 62
AK049743.1 - 6 - 7 624 62
AK146791.1 - 6 - 7 620 62
AK147103.1 - 6 - 7 611 62
AK147909.1 - 6 - 7 601 62
AK150345.1 - 6 - 7 602 62
AK151625.1 - 6 - 7 615 62
AK153276.1 - 6 - 7 602 62
BC068226.1 - 6 - 7 611 5707017 62
X79233.1 - 6 - 7 633 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ewsr1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
nucleic acid binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0