This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187846_g1
See other TMED4 GE Assays ›
Gene Symbol:
Tmed4
Gene Name:
transmembrane emp24 protein transport domain containing 4
Gene Aliases:
1110014L17Rik, AI326346
Chromosome Location:
Chr.11: 6270714 - 6274837 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187846_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmed4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134020.1 NP_598781.1 2 - 3 263 67
GenBank mRNA AF368921.1 - 2 - 3 294 67
AK078425.1 - 2 - 3 274 67
AK158876.1 - 2 - 3 296 67
BC023041.1 - 2 - 3 263 5360236 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002167.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmed4

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

transport
signal transduction
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
signal transducer activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100021132.mm.1 NM_134020.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021132.mm.1 NM_134020.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021132.mm.1 TC1100002167.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0