This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187877_m1
See other IKZF1 GE Assays ›
Gene Symbol:
Ikzf1
Gene Name:
IKAROS family zinc finger 1
Gene Aliases:
5832432G11Rik, Ikaros, LyF-1, Zfpn1a1, Znfn1a1, hlk-1, mKIAA4227
Chromosome Location:
Chr.11: 11684969 - 11772926 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187877_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ikzf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK036833.1 - 2 - 3 297 73
AK040268.1 - 2 - 3 292 73
AK077815.1 - 2 - 3 595 73
AK142856.1 - 2 - 3 179 73
AK154175.1 - 2 - 3 320 73
AK220235.1 - 2 - 3 264 73
AY536269.1 - 1 - 2 37 73
BC138789.1 - 2 - 3 90 8861811 73
L03547.1 - 2 - 3 75 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000134.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100001332.mm.1 NM_001025597.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100001332.mm.1 NM_009578.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100001332.mm.1 TC1100000134.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100001330.mm.1 NM_001025597.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100001330.mm.1 NM_009578.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100001330.mm.1 TC1100000134.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 304340 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bs78j:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0