This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187880_m1
See other IKZF1 GE Assays ›
Gene Symbol:
Ikzf1
Gene Name:
IKAROS family zinc finger 1
Gene Aliases:
5832432G11Rik, Ikaros, LyF-1, Zfpn1a1, Znfn1a1, hlk-1, mKIAA4227
Chromosome Location:
Chr.11: 11684969 - 11772926 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187880_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ikzf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK036833.1 - 5 - 6 850 115
AK040268.1 - 4 - 5 584 115
AK077815.1 - 5 - 6 1148 115
AK154175.1 - 5 - 6 873 115
AK220235.1 - 5 - 6 817 115
BC138789.1 - 4 - 5 382 8861811 115
L03547.1 - 4 - 5 367 115
S74708.1 - 2 - 3 428 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bs78j:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0