This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188300_g1
See other MUTYH GE Assays ›
Gene Symbol:
Mutyh
Gene Name:
mutY homolog (E. coli)
Gene Aliases:
5730495A01Rik, Mutyha, Mutyhalpha, Mutyhb, Mutyhbeta, Mutyhc, Myh
Chromosome Location:
Chr.4: 116807682 - 116819439 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188300_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mutyh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159581.1 NP_001153053.1 14 - 15 1372 136
NM_133250.2 NP_573513.2 13 - 14 1279 136
GenBank mRNA AB117937.1 - 11 - 12 1167 136
AB117938.1 - 12 - 13 1241 136
AK017730.1 - 12 - 13 1169 136
AK049542.1 - 5 - 6 621 136
AK156790.1 - 14 - 15 1362 136
AY007717.1 - 12 - 13 1235 136
BC057942.1 - 13 - 14 1260 5320744 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001266.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mutyh

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
metabolic process
purine-specific mismatch base pair DNA N-glycosylase activity
catalytic activity
uracil DNA N-glycosylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
MutSalpha complex binding
DNA-3-methylbase glycosylase activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978122 BC057942.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978122 AK077546.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978122 AK156790.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978122 AY007717.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978122 AB117937.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 705797 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464016 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601844 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188512 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538042 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97464 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519433 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452168 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167632 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20588 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166040 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151824 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152448 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978122 BC057942.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978122 AK077546.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978122 AK156790.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978122 AY007717.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978122 AB117937.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P238785 AK077546.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P238785 AK017730.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P257619 AY007717.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165831 BC057942.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165831 CX213088.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165831 AK077546.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165831 AK156790.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P165831 AY007717.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23231101 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23231104 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166040 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152448 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151824 - -
Whole Mouse Genome G4122A A_52_P238785 AK077546.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P238785 AK017730.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P165831 BC057942.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P165831 CX213088.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P165831 AK077546.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P165831 AK156790.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P165831 AY007717.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P257619 AY007717.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119111 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106275 AY007717.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152448 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151824 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166040 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03839694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231108 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231059 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231098 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231111 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231089 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231057 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231069 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231028 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231079 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231144 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231060 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231045 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231023 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231051 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231013 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231104 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231115 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231099 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231096 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231016 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231126 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231151 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231063 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231015 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231044 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231011 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231026 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231048 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231085 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231095 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231073 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231055 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231074 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23231124 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03839692 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231088 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231084 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231098 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231059 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03839697 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04084616 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03230771 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231064 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231089 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231073 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03875546 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04020002 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03230764 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231086 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03230766 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04019988 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03839695 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231096 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231097 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03875550 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231099 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231069 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231057 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03839691 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03839694 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04019986 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231092 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231051 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231063 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231054 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04084624 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231095 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231082 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23231078 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10507369 - -
Affymetrix Mouse Gene 1.0 ST Array 10507373 - -
Affymetrix Mouse Gene 1.0 ST Array 10507363 - -
Affymetrix Mouse Gene 1.0 ST Array 10507378 - -
Affymetrix Mouse Gene 1.0 ST Array 10507366 - -
Affymetrix Mouse Gene 1.0 ST Array 10507364 - -
Affymetrix Mouse Gene 1.0 ST Array 10507377 - -
Affymetrix Mouse Gene 1.0 ST Array 10507375 - -
Affymetrix Mouse Gene 1.0 ST Array 10507365 - -
Affymetrix Mouse Gene 1.0 ST Array 10507376 - -
Affymetrix Mouse Gene 1.0 ST Array 10507370 - -
Affymetrix Mouse Gene 1.0 ST Array 10507374 - -
Affymetrix Mouse Gene 1.0 ST Array 10507368 - -
Affymetrix Mouse Gene 1.0 ST Array 10507372 - -
Affymetrix Mouse Gene 1.0 ST Array 10507371 - -
Affymetrix Mouse Gene 1.0 ST Array 10507367 - -
Affymetrix HT MG-430 PM Array Plate 1422816_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10507362 - -
Mouse Genome 430 2.0 Array 1422816_PM_a_at - -
Mouse Genome 430 2.0 Array 1422816_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 70603_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 70603_at - -
Affymetrix Mouse Clariom S Array PSR0400009487.mm.1 NM_001159581.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009487.mm.1 NM_001159581.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009487.mm.1 NM_133250.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009487.mm.1 NM_133250.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400009487.mm.1 TC0400001266.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400009491.mm.1 NM_001159581.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009491.mm.1 NM_001159581.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009491.mm.1 NM_133250.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009491.mm.1 NM_133250.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400009491.mm.1 TC0400001266.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400009492.mm.1 NM_001159581.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009492.mm.1 NM_001159581.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009492.mm.1 NM_133250.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009492.mm.1 NM_133250.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400009492.mm.1 TC0400001266.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400009489.mm.1 NM_001159581.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009489.mm.1 NM_001159581.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009489.mm.1 NM_133250.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009489.mm.1 NM_133250.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400009489.mm.1 TC0400001266.mm.1 MTA TC
Mouse Expression Set 430 1422816_a_at - -
Mouse Expression Set 430 1422816_PM_a_at - -
Mouse Genome 430A 2.0 Array 1422816_a_at - -
Mouse Genome 430A 2.0 Array 1422816_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1422816_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1422816_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 471721 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102650520 - -
MouseWG-6 v2 Expression BeadChip 1780441 - -
Illumina mouse-6 v1.0 expression beadchip 1780441 - -
Illumina mouse-6 v1.0 expression beadchip 102650520 - -
Illumina mouse-6 v1.0 expression beadchip 5700121 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001780441 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005700121 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220545 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2590061 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1220545 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1219741 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2590061 - -
mouse-6 v1.1 (Illumina) scl25062.15.1_98-S - -
mouse-6 v1.1 (Illumina) scl0070603.2_90-S - -
mouse-6 v1.1 (Illumina) scl0002693.1_3-S - -
Illumina Sentrix mouse-6.v1 bead array 5700121 - -
Illumina Sentrix mouse-6.v1 bead array 102650520 - -
Illumina Sentrix mouse-6.v1 bead array 1780441 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0002693.1_3-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0070603.2_90-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780441 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5700121 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102650520 - -
MouseRef-8 v2 Expression BeadChip ILMN_2590061 - -
MouseRef-8 v2 Expression BeadChip ILMN_1220545 - -
MouseRef-8 v2 Expression BeadChip ILMN_2920924 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2590061 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220545 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2920924 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0