| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400009475.mm.1 |
NM_001159581.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009475.mm.1 |
NM_001159581.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009475.mm.1 |
NM_133250.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009475.mm.1 |
NM_133250.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009475.mm.1 |
TC0400001266.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009473.mm.1 |
NM_001159581.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009473.mm.1 |
NM_001159581.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009473.mm.1 |
NM_133250.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009473.mm.1 |
NM_133250.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009473.mm.1 |
TC0400001266.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009474.mm.1 |
NM_001159581.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009474.mm.1 |
NM_001159581.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009474.mm.1 |
NM_133250.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009474.mm.1 |
NM_133250.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009474.mm.1 |
TC0400001266.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400009477.mm.1 |
NM_001159581.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009477.mm.1 |
NM_001159581.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009477.mm.1 |
NM_133250.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009477.mm.1 |
NM_133250.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400009477.mm.1 |
TC0400001266.mm.1 |
MTA TC |