This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188315_g1
See other ELOVL1 GE Assays ›
Gene Symbol:
Elovl1
Gene Name:
elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 1
Gene Aliases:
AA407424, BB151133, Ssc1
Chromosome Location:
Chr.4: 118428081 - 118432953 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188315_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elovl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039175.2 NP_001034264.1 3 - 4 262 83
NM_001039176.2 NP_001034265.1 4 - 5 460 83
NM_019422.3 NP_062295.1 4 - 5 453 83
GenBank mRNA AF170907.1 - 4 - 5 446 83
AK003743.1 - 3 - 4 231 83
AK135348.1 - 4 - 5 428 83
AK156003.1 - 4 - 5 2054 83
AK171217.1 - 4 - 5 439 83
AK205597.1 - 2 - 3 254 83
BC006735.1 - 4 - 5 460 3156004 83
BC096673.1 - 3 - 4 241 5143739 83
CN455861.1 - 2 - 3 197 83
CX199666.1 - 4 - 5 426 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001307.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Elovl1

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
fatty acid elongation, saturated fatty acid
sphingolipid biosynthetic process
fatty acid elongation, monounsaturated fatty acid
very long-chain fatty acid biosynthetic process
fatty acid elongase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lz5mk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0