This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188467_g1
See other GALE GE Assays ›
Gene Symbol:
Gale
Gene Name:
galactose-4-epimerase, UDP
Gene Aliases:
2310002A12Rik, AI323962
Chromosome Location:
Chr.4: 135963681 - 135968178 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188467_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gale
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178389.3 NP_848476.1 5 - 6 810 103
GenBank mRNA AK009083.1 - 6 - 7 678 103
BC027438.1 - 5 - 6 810 4224021 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001671.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gale

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
galactose metabolic process
galactose catabolic process
catalytic activity
UDP-N-acetylglucosamine 4-epimerase activity
UDP-glucose 4-epimerase activity
isomerase activity
racemase and epimerase activity, acting on carbohydrates and derivatives
protein homodimerization activity
coenzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400013400.mm.1 NM_178389.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400013400.mm.1 NM_178389.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400013400.mm.1 TC0400001671.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f9n9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0