This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188585_m1
See other HYI GE Assays ›
Gene Symbol:
Hyi
Gene Name:
hydroxypyruvate isomerase homolog (E. coli)
Gene Aliases:
2700033B16Rik, 6430559E15Rik, HT036
Chromosome Location:
Chr.4: 118359999 - 118362744 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188585_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hyi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003946.2 1 - 2 237 123
GenBank mRNA AK020099.1 - 1 - 2 223 123
CK620463.1 - 1 - 2 237 123
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA018004.1 - 1 - 2 163 Chr. 4 - 118360071 - 118362073 [+] on Build GRCm38 123
GenBankmRNA AA277622.1 - 1 - 2 168 Chr. 4 - 118360073 - 118362246 [+] on Build GRCm38 123
GenBankmRNA GH454744.1 - 266 Chr. - - [-] on Build 123
GenBankmRNA W83198.1 - 154 Chr. - - [-] on Build 123
GenBankmRNA BY719057.1 - 1 - 2 217 Chr. 4 - 118360019 - 118362497 [+] on Build GRCm38 123

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein tyrosine phosphatase,
receptor type,
F
Ptprf AA591035,
LAR,
LARS,
RPTP-LAR
-68.6kb Anti-sense Chr. 4 - 118208213 - 118291400 on NCBI Build GRCm38
Downstream seizure threshold 2 Szt2 6430407N22,
ENSMUSG00000058432,
TIGR
1.0kb Anti-sense Chr. 4 - 118362740 - 118409276 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001305.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
hydroxypyruvate isomerase activity
isomerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400009783.mm.1 NR_003946.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009783.mm.1 NR_003946.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400009783.mm.1 TC0400001305.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400009789.mm.1 NR_003946.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400009789.mm.1 NR_003946.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400009789.mm.1 TC0400001305.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline