This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188755_m1
See other CNBP GE Assays ›
Gene Symbol:
Cnbp
Gene Name:
cellular nucleic acid binding protein
Gene Aliases:
AA408710, Cnbp1, Znf9
Chromosome Location:
Chr.6: 87842615 - 87851106 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188755_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnbp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109745.1 NP_001103215.1 1 - 2 206 58
NM_001109746.1 NP_001103216.1 1 - 2 206 58
NM_013493.3 NP_038521.1 1 - 2 206 58
GenBank mRNA AK075760.1 - 1 - 2 171 58
AK078427.1 - 1 - 2 88 58
AK144740.1 - 1 - 2 153 58
AK145444.1 - 1 - 2 136 58
AK146224.1 - 1 - 2 147 58
AK146301.1 - 1 - 2 149 58
AK146344.1 - 1 - 2 149 58
AK146593.1 - 1 - 2 122 58
AK147157.1 - 1 - 2 131 58
AK151317.1 - 1 - 2 149 58
AK151528.1 - 1 - 2 119 58
AK151961.1 - 1 - 2 147 58
AK153414.1 - 1 - 2 125 58
AK159810.1 - 1 - 2 123 58
BC058723.1 - 1 - 2 95 5705640 58
CJ088264.1 - 1 - 2 206 58
L12693.1 - 1 - 2 93 58
U20326.1 - 1 - 2 108 58
Z11871.1 - 1 - 2 93 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK075760.1, AK078427.1, AK144740.1, AK145444.1, AK146224.1, AK146301.1, AK146344.1, AK146593.1, AK151317.1, AK151528.1, AK151961.1, AK153414.1, AK159810.1, BC058723.1, L12693.1, NM_001109745.1, NM_001109746.1, NM_013493.3, U20326.1, Z11871.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cnbp

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell proliferation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nucleic acid binding
DNA binding
single-stranded DNA binding
single-stranded RNA binding
zinc ion binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mfdv2:80/100.66.77.127:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0