This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188799_m1
See other APOBEC1 GE Assays ›
Gene Symbol:
Apobec1
Gene Name:
apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1
Gene Aliases:
Cdar1
Chromosome Location:
Chr.6: 122577792 - 122603041 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188799_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apobec1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA U22262.1 - 2 - 3 257 61
U22263.1 - 2 - 3 257 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the U22262.1, U22263.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 911726 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0