Assay Details
| Gene Symbol: Rab43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133717.3 | NP_598478.1 | 1 - 2 | 132 | 74 | |
| GenBank mRNA | BC010248.1 | - | 1 - 2 | 132 | 3155089 | 74 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC010248.1, NM_133717.3 transcripts.
Set Membership: |
|

Rab43
Genomic Map