This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01189292_m1
See other DSE GE Assays ›
Gene Symbol:
Dse
Gene Name:
dermatan sulfate epimerase
Gene Aliases:
6030499O08, AI480506, B130024B19Rik, DS-epi1, Sart2
Chromosome Location:
Chr.10: 34151386 - 34208275 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01189292_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dse
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172508.2 NP_766096.1 3 - 4 1189 67
GenBank mRNA AK031737.1 - 3 - 4 876 67
AK045065.1 - 3 - 4 1096 67
AK150737.1 - 3 - 4 1353 67
AK153326.1 - 3 - 4 862 67
BC053095.1 - 3 - 4 1189 6401066 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001995.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dse

Gene Ontology Categories:

Function(s) Process(es)

dermatan sulfate biosynthetic process
isomerase activity
chondroitin-glucuronate 5-epimerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000014206.mm.1 NM_172508.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014206.mm.1 NM_172508.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000014206.mm.1 TC1000001995.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0