This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01189293_m1
See other DSE GE Assays ›
Gene Symbol:
Dse
Gene Name:
dermatan sulfate epimerase
Gene Aliases:
6030499O08, AI480506, B130024B19Rik, DS-epi1, Sart2
Chromosome Location:
Chr.10: 34151386 - 34208275 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01189293_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dse
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172508.2 NP_766096.1 4 - 5 1438 84
GenBank mRNA AK031737.1 - 4 - 5 1125 84
AK045065.1 - 4 - 5 1345 84
AK150737.1 - 4 - 5 1602 84
AK153326.1 - 4 - 5 1111 84
BC053095.1 - 4 - 5 1438 6401066 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001995.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dse

Gene Ontology Categories:

Function(s) Process(es)

dermatan sulfate biosynthetic process
isomerase activity
chondroitin-glucuronate 5-epimerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000014205.mm.1 NM_172508.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014205.mm.1 NM_172508.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000014205.mm.1 TC1000001995.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f9n9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0