This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01189491_g1
See other IL17RE GE Assays ›
Gene Symbol:
Il17re
Gene Name:
interleukin 17 receptor E
Gene Aliases:
AA589509, IL-17RE, Il25r
Chromosome Location:
Chr.6: 113458484 - 113470758 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01189491_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il17re
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034029.1 NP_001029201.1 8 - 9 884 67
NM_001034031.1 NP_001029203.1 10 - 11 959 67
NM_145826.5 NP_665825.2 9 - 10 1311 67
GenBank mRNA AF458068.1 - 9 - 10 961 67
AK046302.1 - 9 - 10 1314 67
AK046713.1 - 9 - 10 973 67
AK078876.1 - 2 - 3 1127 67
AK169498.1 - 9 - 10 1139 67
BC069861.1 - 9 - 10 988 6311568 67
DQ092339.1 - 9 - 10 900 67
DQ092340.1 - 8 - 9 884 67
DQ092341.1 - 10 - 11 959 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK078876.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il17re

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cytokine-mediated signaling pathway
interleukin-17 receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600009749.mm.1 TC0600001251.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600009749.mm.1 NM_001034029.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009749.mm.1 NM_001034029.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009749.mm.1 NM_001034031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009749.mm.1 NM_001034031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009749.mm.1 NM_145826.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009749.mm.1 NM_145826.5 Refseq
Affymetrix Mouse Clariom S Array PSR0600009748.mm.1 TC0600001251.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600009748.mm.1 NM_001034029.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009748.mm.1 NM_001034029.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009748.mm.1 NM_001034031.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009748.mm.1 NM_001034031.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009748.mm.1 NM_145826.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009748.mm.1 NM_145826.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0