This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01189576_m1
See other PPIL6 GE Assays ›
Gene Symbol:
Ppil6
Gene Name:
peptidylprolyl isomerase (cyclophilin)-like 6
Gene Aliases:
2900084F20Rik
Chromosome Location:
Chr.10: 41490439 - 41514295 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01189576_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppil6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028430.1 NP_082706.1 1 - 2 184 64
GenBank mRNA AK013818.1 - 1 - 2 147 64
BC147756.1 - 1 - 2 180 9056705 64
BC147757.1 - 1 - 2 180 9056706 64
BY104076.1 - 1 - 2 185 64
CA466046.1 - 1 - 2 166 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000366.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
biological_process
molecular_function
peptidyl-prolyl cis-trans isomerase activity
isomerase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 NM_028430.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 AI463820.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 AK013818.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 BC147756.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 BC147757.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 NM_028430.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 AI463820.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 AK013818.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 BC147756.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224042 BC147757.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925143 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628608 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204744 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540282 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372303 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491081 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683295 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181968 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762102 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813913 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207137 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957647 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942339 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853531 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81658 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143348 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176514 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135441 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 NM_028430.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 AI463820.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 AK013818.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 BC147756.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 BC147757.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 NM_028430.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 AI463820.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 AK013818.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 BC147756.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224042 BC147757.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 NM_028430.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 AI463820.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 AK013818.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 BC147756.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 BC147757.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 NM_028430.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 AI463820.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 AK013818.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 BC147756.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P224042 BC147757.1 mouse_transcriptome
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089727 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089739 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089556 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089704 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089723 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089568 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089735 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089681 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089662 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089558 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089605 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089718 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089747 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089717 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089592 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089729 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089742 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089555 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089713 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089711 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089684 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089685 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089737 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089678 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089756 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089758 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089732 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089725 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089561 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089698 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089674 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089720 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27089576 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089685 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089665 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089684 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089579 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08523387 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089578 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089600 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08427823 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089680 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089682 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089681 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089584 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089669 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089668 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08947534 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089662 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089574 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089585 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089569 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08863999 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08354097 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089575 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089590 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089572 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089660 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089556 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08286500 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089565 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08620146 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089564 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089597 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089688 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089566 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089568 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089561 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089570 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089563 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089583 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08620153 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089594 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089655 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089602 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27089576 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10362797 - -
Affymetrix Mouse Gene 1.0 ST Array 10362796 - -
Affymetrix Mouse Gene 1.0 ST Array 10362800 - -
Affymetrix Mouse Gene 1.0 ST Array 10362801 - -
Affymetrix Mouse Gene 1.0 ST Array 10362802 - -
Affymetrix Mouse Gene 1.0 ST Array 10362799 - -
Affymetrix Mouse Gene 1.0 ST Array 10362798 - -
Affymetrix HT MG-430 PM Array Plate 1453310_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10362794 - -
Mouse Genome 430 2.0 Array 1453310_at - -
Mouse Genome 430 2.0 Array 1453310_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 73075_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 73075_PM_at - -
Affymetrix Mouse Clariom S Array PSR1000002524.mm.1 NM_028430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002524.mm.1 NM_028430.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002524.mm.1 TC1000000366.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000002523.mm.1 NM_028430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002523.mm.1 NM_028430.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002523.mm.1 TC1000000366.mm.1 MTA TC
Mouse Expression Set 430 1453310_PM_at - -
Mouse Expression Set 430 1453310_at - -
Murine Genome U74v2 Set 166791_PM_at - -
Murine Genome U74v2 Set 166791_at - -
Murine Genome U74v2 Set 169115_PM_i_at - -
Murine Genome U74v2 Set 169115_i_at - -
Affymetrix HT Mouse Genome 430B Array 1453310_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453310_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 811330 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2481541 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2481541 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0