This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01189580_m1
See other PPIL6 GE Assays ›
Gene Symbol:
Ppil6
Gene Name:
peptidylprolyl isomerase (cyclophilin)-like 6
Gene Aliases:
2900084F20Rik
Chromosome Location:
Chr.10: 41490439 - 41514295 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01189580_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppil6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028430.1 NP_082706.1 5 - 6 672 95
GenBank mRNA AI463820.1 - 3 - 4 307 95
AK013818.1 - 5 - 6 635 95
BC147756.1 - 5 - 6 668 9056705 95
BC147757.1 - 5 - 6 668 9056706 95
BP772786.1 - 3 - 4 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000366.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
biological_process
molecular_function
peptidyl-prolyl cis-trans isomerase activity
isomerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000002528.mm.1 NM_028430.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002528.mm.1 NM_028430.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002528.mm.1 TC1000000366.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0