This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01190075_m1
See other ARHGAP25 GE Assays ›
Gene Symbol:
Arhgap25
Gene Name:
Rho GTPase activating protein 25
Gene Aliases:
A130039I20Rik
Chromosome Location:
Chr.6: 87458544 - 87533259 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01190075_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286610.1 NP_001273539.1 1 - 2 434 95
GenBank mRNA AK159598.1 - 1 - 2 435 95
AK159941.1 - 1 - 2 435 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK159598.1, AK159941.1, NM_001286610.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600020655.mm.1 TC0600002656.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600020655.mm.1 NM_001037727.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600020655.mm.1 NM_001286610.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020649.mm.1 TC0600002656.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600020649.mm.1 NM_001037727.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600020649.mm.1 NM_001286610.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020649.mm.1 NM_175476.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600020649.mm.1 NR_104483.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0