Assay Details
| Gene Symbol: Hace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172473.3 | NP_766061.2 | 3 - 4 | 467 | 66 | |
| GenBank mRNA | AK042879.1 | - | 3 - 4 | 408 | 66 | |
| AK156958.1 | - | 3 - 4 | 409 | 66 | ||
| BC120695.1 | - | 3 - 4 | 364 | 40129618 | 66 | |
| BC120697.1 | - | 3 - 4 | 364 | 40129620 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hace1
Genomic Map