This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01190692_g1
See other HACE1 GE Assays ›
Gene Symbol:
Hace1
Gene Name:
HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1
Gene Aliases:
1700042J16Rik, A730034A22Rik, BC025474
Chromosome Location:
Chr.10: 45577571 - 45712345 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01190692_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hace1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172473.3 NP_766061.2 8 - 9 952 139
GenBank mRNA AK042879.1 - 8 - 9 893 139
AK156958.1 - 8 - 9 894 139
BC120695.1 - 8 - 9 849 40129618 139
BC120697.1 - 8 - 9 849 40129620 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000430.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hace1

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
Golgi organization
cell cycle
protein ubiquitination
regulation of cell migration
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
membrane fusion
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
Rab GTPase binding
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
Rac GTPase binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000002901.mm.1 NM_172473.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002901.mm.1 NM_172473.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000002901.mm.1 TC1000000430.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000002902.mm.1 NM_172473.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002902.mm.1 NM_172473.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000002902.mm.1 TC1000000430.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline