This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01190797_m1
See other ASF1A GE Assays ›
Gene Symbol:
Asf1a
Gene Name:
anti-silencing function 1A histone chaperone
Gene Aliases:
2310079C17Rik, C79052
Chromosome Location:
Chr.10: 53596961 - 53609225 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01190797_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asf1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025541.3 NP_079817.1 1 - 2 345 78
GenBank mRNA AK007804.1 - 1 - 2 201 78
AK010210.1 - 1 - 2 355 78
AK012376.1 - 1 - 2 336 78
AK031513.1 - 1 - 2 311 78
AK088618.1 - 1 - 2 288 78
BC027628.1 - 1 - 2 137 1178866 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000484.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asf1a

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
DNA repair
chromatin assembly or disassembly
nucleosome assembly
DNA replication-dependent nucleosome assembly
DNA replication-independent nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
muscle cell differentiation
chromatin binding
histone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000003189.mm.1 NM_025541.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003189.mm.1 NM_025541.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003189.mm.1 TC1000000484.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000003190.mm.1 NM_025541.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003190.mm.1 NM_025541.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003190.mm.1 TC1000000484.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000003188.mm.1 NM_025541.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003188.mm.1 NM_025541.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003188.mm.1 TC1000000484.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0