This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191514_g1
See other INPP5J GE Assays ›
Gene Symbol:
Inpp5j
Gene Name:
inositol polyphosphate 5-phosphatase J
Gene Aliases:
Pib5pa, Pipp
Chromosome Location:
Chr.11: 3494272 - 3505567 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191514_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Inpp5j
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172439.3 NP_766027.2 12 - 13 2579 63
GenBank mRNA AK034272.1 - 12 - 13 2580 63
AK136908.1 - 12 - 13 2567 63
AK170114.1 - 12 - 13 2545 63
AK171029.1 - 12 - 13 2540 63
BC051452.1 - 12 - 13 2548 63
BC131634.1 - 12 - 13 2574 40096940 63
BC131635.1 - 12 - 13 2571 40096943 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002091.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Inpp5j

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of neuron projection development
dephosphorylation
negative regulation of microtubule polymerization
negative regulation of peptidyl-serine phosphorylation
phosphatidylinositol dephosphorylation
inositol-polyphosphate 5-phosphatase activity
hydrolase activity
SH3 domain binding
phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
inositol-1,4,5-trisphosphate 5-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115871 BY629401.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115871 BC046561.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115871 BC051452.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115871 BC022915.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115871 AK034272.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15626 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579894 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790756 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895777 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413059 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94110 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153777 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133805 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115871 BY629401.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115871 BC046561.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115871 BC051452.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115871 BC022915.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115871 AK034272.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339912 BC131634.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339912 BC131635.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339912 BC046561.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339912 BC051452.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339912 AK171029.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552344 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552353 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552370 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552373 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552377 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552410 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552401 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552351 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552385 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552345 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552355 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552384 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552379 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552338 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552366 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552400 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552363 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552371 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552383 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552350 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552378 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552357 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552365 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552391 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552387 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552393 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552349 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552397 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552375 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552368 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552388 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552361 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552339 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552341 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552347 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552409 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552343 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552404 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552406 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552376 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552394 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552381 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552403 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552360 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27552369 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07238317 - -
Whole Mouse Genome G4122A A_52_P339912 BC131634.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P339912 BC131635.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P339912 BC046561.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P339912 BC051452.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P339912 AK171029.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116997 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133660 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09407221 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09375904 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09038897 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09375911 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09375903 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09375909 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09468897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552239 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552248 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552241 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552252 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552235 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552225 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552242 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552258 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552320 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552331 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552246 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552270 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27552301 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552354 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09118927 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552307 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552349 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09362676 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552331 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09375904 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552381 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552365 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552347 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09118938 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552383 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552372 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552353 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09483758 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552326 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09375911 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552351 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552343 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552366 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552360 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09118925 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09034939 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09608964 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552311 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552375 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09226732 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552344 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552368 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552288 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552350 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552296 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552315 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09118929 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09437821 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09118937 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552290 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552322 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09529929 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552301 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552325 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552294 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09181066 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552333 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09118942 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09181072 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552297 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552320 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552345 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552339 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552335 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552357 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552369 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09034941 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09181073 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552324 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552309 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27552370 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10383719 - -
Affymetrix Mouse Gene 1.0 ST Array 10383720 - -
Affymetrix Mouse Gene 1.0 ST Array 10383723 - -
Affymetrix Mouse Gene 1.0 ST Array 10383727 - -
Affymetrix Mouse Gene 1.0 ST Array 10383725 - -
Affymetrix Mouse Gene 1.0 ST Array 10383724 - -
Affymetrix Mouse Gene 1.0 ST Array 10383721 - -
Affymetrix Mouse Gene 1.0 ST Array 10383718 - -
Affymetrix Mouse Gene 1.0 ST Array 10383728 - -
Affymetrix Mouse Gene 1.0 ST Array 10383722 - -
Affymetrix Mouse Gene 1.0 ST Array 10383726 - -
Affymetrix Mouse Gene 1.0 ST Array 10383730 - -
Affymetrix Mouse Gene 1.0 ST Array 10383729 - -
Affymetrix HT MG-430 PM Array Plate 1433941_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10383717 - -
Mouse Genome 430 2.0 Array 1433941_at - -
Mouse Genome 430 2.0 Array 1441247_PM_at - -
Mouse Genome 430 2.0 Array 1441247_at - -
Mouse Genome 430 2.0 Array 1433941_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 170835_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 170835_at - -
Affymetrix Mouse Clariom S Array PSR1100020245.mm.1 NM_172439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020245.mm.1 NM_172439.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100020245.mm.1 TC1100002091.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100020243.mm.1 NM_172439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020243.mm.1 NM_172439.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100020243.mm.1 TC1100002091.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100020240.mm.1 NM_172439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020240.mm.1 NM_172439.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100020240.mm.1 TC1100002091.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100020242.mm.1 NM_172439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020242.mm.1 NM_172439.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100020242.mm.1 TC1100002091.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100020244.mm.1 NM_172439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020244.mm.1 NM_172439.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100020244.mm.1 TC1100002091.mm.1 MTA TC
Mouse Expression Set 430 1433941_at - -
Mouse Expression Set 430 1433941_PM_at - -
Murine Genome U74v2 Set 103732_at - -
Murine Genome U74v2 Set 103732_PM_at - -
Mouse Genome 430A 2.0 Array 1433941_at - -
Mouse Genome 430A 2.0 Array 1433941_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1433941_at - -
Affymetrix HT Mouse Genome 430A Array 1433941_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6900427 - -
Illumina mouse-6 v1.0 expression beadchip 6900427 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006900427 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228073 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1228073 - -
mouse-6 v1.1 (Illumina) scl40598.13.96_215-S - -
Illumina Sentrix mouse-6.v1 bead array 6900427 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl40598.13.96_215-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6900427 - -
MouseRef-8 v2 Expression BeadChip ILMN_3008586 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3008586 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40598.13.96_215-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline