This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191515_g1
See other INPP5J GE Assays ›
Gene Symbol:
Inpp5j
Gene Name:
inositol polyphosphate 5-phosphatase J
Gene Aliases:
Pib5pa, Pipp
Chromosome Location:
Chr.11: 3494272 - 3505567 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191515_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Inpp5j
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172439.3 NP_766027.2 3 - 4 1451 69
GenBank mRNA AK034272.1 - 3 - 4 1452 69
AK136908.1 - 3 - 4 1438 69
AK165718.1 - 3 - 4 1822 69
AK170114.1 - 3 - 4 1417 69
BC022915.1 - 3 - 4 1404 69
BC046561.1 - 2 - 3 264 69
BC051452.1 - 3 - 4 1421 69
BC131634.1 - 3 - 4 1446 40096940 69
BC131635.1 - 3 - 4 1443 40096943 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002091.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Inpp5j

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of neuron projection development
dephosphorylation
negative regulation of microtubule polymerization
negative regulation of peptidyl-serine phosphorylation
phosphatidylinositol dephosphorylation
inositol-polyphosphate 5-phosphatase activity
hydrolase activity
SH3 domain binding
phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
inositol-1,4,5-trisphosphate 5-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100020257.mm.1 NM_172439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020257.mm.1 NM_172439.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100020257.mm.1 TC1100002091.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100020255.mm.1 NM_172439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020255.mm.1 NM_172439.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100020255.mm.1 TC1100002091.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline