This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191812_m1
See other AKR1A1 GE Assays ›
Gene Symbol:
Akr1a1
Gene Name:
aldo-keto reductase family 1, member A1 (aldehyde reductase)
Gene Aliases:
2610201A18Rik, Akr1a4
Chromosome Location:
Chr.4: 116636510 - 116651674 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191812_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Akr1a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021473.3 NP_067448.1 1 - 2 334 94
GenBank mRNA AF225564.1 - 1 - 2 239 94
AK005376.1 - 1 - 2 270 94
AK009462.1 - 1 - 2 111 94
AK011321.1 - 1 - 2 263 94
AK011388.1 - 1 - 2 289 94
AK011667.1 - 1 - 2 290 94
AK011794.1 - 1 - 2 263 94
AK011856.1 - 1 - 2 290 94
AK011906.1 - 1 - 2 268 94
AK011908.1 - 1 - 2 303 94
AK011918.1 - 1 - 2 289 94
AK128906.1 - 1 - 2 56 94
AK144198.1 - 1 - 2 290 94
AK146081.1 - 1 - 2 288 94
AK146278.1 - 1 - 2 290 94
AK152086.1 - 1 - 2 269 94
AK159573.1 - 1 - 2 288 94
AK164461.1 - 1 - 2 289 94
AK168950.1 - 1 - 2 285 94
AK186333.1 - 1 - 2 91 94
AK192030.1 - 1 - 2 92 94
AK210646.1 - 1 - 2 91 94
AY207463.1 - 1 - 2 311 94
BC003850.1 - 1 - 2 118 94
BC039926.1 - 1 - 2 283 3156658 94
BC046762.1 - 1 - 2 240 5693736 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF225564.1, AK009462.1, AK011321.1, AK011388.1, AK011667.1, AK011794.1, AK011856.1, AK011906.1, AK011908.1, AK011918.1, AK144198.1, AK146081.1, AK146278.1, AK152086.1, AK164461.1, AK168950.1, AY207463.1, BC039926.1, BC046762.1, NM_021473.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Akr1a1

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

L-ascorbic acid biosynthetic process
D-glucuronate catabolic process
aldehyde catabolic process
oxidation-reduction process
alditol:NADP+ 1-oxidoreductase activity
alcohol dehydrogenase (NADP+) activity
oxidoreductase activity
L-glucuronate reductase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400026759.mm.1 TC0400003344.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026759.mm.1 NM_021473.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026759.mm.1 NM_021473.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400026757.mm.1 TC0400003344.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026757.mm.1 NM_021473.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026757.mm.1 NM_021473.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400026758.mm.1 TC0400003344.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026758.mm.1 NM_021473.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026758.mm.1 NM_021473.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400026754.mm.1 TC0400003344.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026754.mm.1 NM_021473.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026754.mm.1 NM_021473.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400026756.mm.1 TC0400003344.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026756.mm.1 NM_021473.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026756.mm.1 NM_021473.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 919925 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0