This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191970_g1
See other NPC1L1 GE Assays ›
Gene Symbol:
Npc1l1
Gene Name:
NPC1-like 1
Gene Aliases:
9130221N23Rik, Gm243
Chromosome Location:
Chr.11: 6211011 - 6230245 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191970_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npc1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207242.2 NP_997125.2 17 - 18 3757 77
GenBank mRNA AK078947.1 - 17 - 18 3658 77
AY437866.1 - 17 - 18 3635 77
BC131788.1 - 17 - 18 3634 77
BC131789.1 - 9 - 10 2599 40086103 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002165.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Npc1l1

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
cholesterol biosynthetic process
steroid metabolic process
cholesterol metabolic process
intestinal cholesterol absorption
cholesterol transport
lipoprotein metabolic process
cholesterol homeostasis
drug binding
hedgehog receptor activity
Rab GTPase binding
myosin V binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100021073.mm.1 NM_207242.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021073.mm.1 NM_207242.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100021073.mm.1 TC1100002165.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0