This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01192214_g1
See other MFSD2A GE Assays ›
Gene Symbol:
Mfsd2a
Gene Name:
major facilitator superfamily domain containing 2A
Gene Aliases:
1700018O18Rik, Mfsd2, NLS1
Chromosome Location:
Chr.4: 122946850 - 122961195 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01192214_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mfsd2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029662.2 NP_083938.2 6 - 7 885 102
GenBank mRNA AK006096.1 - 6 - 7 884 102
BC046793.1 - 6 - 7 867 5717701 102
BC060526.1 - 6 - 7 856 5707105 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003475.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mfsd2a

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
fatty acid transport
phospholipid transport
transcytosis
transmembrane transport
establishment of blood-brain barrier
lipid transport across blood brain barrier
phospholipid transporter activity
symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400028160.mm.1 TC0400003475.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400028160.mm.1 NM_029662.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028160.mm.1 NM_029662.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0