This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01192244_m1
See other PPIE GE Assays ›
Gene Symbol:
Ppie
Gene Name:
peptidylprolyl isomerase E (cyclophilin E)
Gene Aliases:
2010010D16Rik, Cyp33
Chromosome Location:
Chr.4: 123127114 - 123140000 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01192244_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppie
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019489.5 NP_062362.1 4 - 5 205 61
GenBank mRNA AK008177.1 - 4 - 5 206 61
BC045154.1 - 4 - 5 200 6468589 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003478.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppie

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
positive regulation of viral genome replication
nucleotide binding
nucleic acid binding
RNA binding
peptidyl-prolyl cis-trans isomerase activity
isomerase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400028186.mm.1 TC0400003478.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400028186.mm.1 NM_019489.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028186.mm.1 NM_019489.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0