This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01192899_m1
See other GPRC6A GE Assays ›
Gene Symbol:
Gprc6a
Gene Name:
G protein-coupled receptor, family C, group 6, member A
Gene Aliases:
-
Chromosome Location:
Chr.10: 51614688 - 51631614 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01192899_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gprc6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153071.1 NP_694711.1 4 - 5 1574 82
GenBank mRNA AY101365.1 - 4 - 5 1574 82
AY735399.1 - 4 - 5 1547 82
AY735400.1 - 4 - 5 1022 82
BC108998.2 - 4 - 5 1574 40048019 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002148.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gprc6a

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
calcium-mediated signaling
response to amino acid
signal transducer activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000015056.mm.1 NM_153071.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015056.mm.1 NM_153071.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015056.mm.1 TC1000002148.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0