This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01192917_m1
See other TBC1D32 GE Assays ›
Gene Symbol:
Tbc1d32
Gene Name:
TBC1 domain family, member 32
Gene Aliases:
Bromi, D630037F22Rik, b2b2284Clo
Chromosome Location:
Chr.10: 56014293 - 56229456 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01192917_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tbc1d32
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033385.3 NP_001028557.2 27 - 28 3154 91
GenBank mRNA AK035346.1 - 9 - 10 934 91
AK141206.1 - 12 - 13 1349 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lens development in camera-type eye
retinal pigment epithelium development
smoothened signaling pathway
multicellular organism development
neural tube development
axoneme assembly
embryonic digit morphogenesis
camera-type eye development
retina development in camera-type eye
cilium morphogenesis
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
protein localization to cilium
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline