This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01193610_m1
See other L1CAM GE Assays ›
Gene Symbol:
L1cam
Gene Name:
L1 cell adhesion molecule
Gene Aliases:
CD171, L1, N-CAM-L1, NCAM-L1
Chromosome Location:
Chr.X: 73853780 - 73880841 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01193610_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: L1cam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008478.3 NP_032504.3 1 - 2 141 77
GenBank mRNA AK139407.1 - 1 - 2 104 77
BC056988.1 - 1 - 2 66 6822217 77
CJ109973.1 - 1 - 2 141 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC056988.1, NM_008478.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene L1cam

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
leukocyte cell-cell adhesion
cell surface receptor signaling pathway
multicellular organism development
nervous system development
axon guidance
positive regulation of cell-cell adhesion
cell differentiation
neuron projection development
cell-cell adhesion mediated by integrin
homotypic cell-cell adhesion
positive regulation of calcium-mediated signaling
integrin binding
protein binding
PDZ domain binding
sialic acid binding
identical protein binding
protein self-association

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0