Assay Details
| Gene Symbol: Ssr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166480.1 | NP_001159952.1 | 4 - 5 | 564 | 128 | |
| NM_009279.3 | NP_033305.1 | 4 - 5 | 446 | 128 | ||
| GenBank mRNA | AK007921.1 | - | 4 - 5 | 402 | 128 | |
| AK013723.1 | - | 4 - 5 | 408 | 128 | ||
| AK160089.1 | - | 4 - 5 | 412 | 128 | ||
| BC008172.1 | - | 4 - 5 | 367 | 3481948 | 128 | |
| BC019419.1 | - | 3 - 4 | 1897 | 128 | ||
| BQ747160.1 | - | 4 - 5 | 349 | 128 | ||
| X90582.1 | - | 4 - 5 | 527 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssr4
Genomic Map