This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01193791_m1
See other VTA1 GE Assays ›
Gene Symbol:
Vta1
Gene Name:
Vps20-associated 1 homolog (S. cerevisiae)
Gene Aliases:
1110001D18Rik, 1110059P08Rik, AU040813, C85340, LIP5, SBP1
Chromosome Location:
Chr.10: 14654760 - 14705527 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01193791_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vta1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025418.3 NP_079694.2 1 - 2 138 79
GenBank mRNA AK002611.1 - 1 - 2 137 79
AK004323.1 - 1 - 2 210 79
AK009227.2 - 1 - 2 127 79
BC026752.1 - 1 - 2 131 4034998 79
CX212626.1 - 1 - 2 139 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vta1

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
viral budding
multivesicular body sorting pathway
protein binding
protein C-terminus binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0