This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194017_m1
See other NDUFB8 GE Assays ›
Gene Symbol:
Ndufb8
Gene Name:
NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8
Gene Aliases:
2900010I05Rik, AI987932
Chromosome Location:
Chr.19: 44548576 - 44555446 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194017_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufb8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026061.2 NP_080337.1 3 - 4 330 73
GenBank mRNA AK013516.1 - 3 - 4 330 73
AK028070.1 - 3 - 4 341 73
AK212374.1 - 1 - 2 40 73
BC043019.1 - 3 - 4 330 5684256 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001777.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufb8

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
transport
biological_process
oxidation-reduction process
molecular_function
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900013888.mm.1 NM_026061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013888.mm.1 NM_026061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013888.mm.1 TC1900001777.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013889.mm.1 NM_026061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013889.mm.1 NM_026061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013889.mm.1 TC1900001777.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013891.mm.1 NM_026061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013891.mm.1 NM_026061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013891.mm.1 TC1900001777.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013887.mm.1 NM_026061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013887.mm.1 NM_026061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013887.mm.1 TC1900001777.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013884.mm.1 NM_026061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013884.mm.1 NM_026061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013884.mm.1 TC1900001777.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900015582.mm.1 NM_026061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900015582.mm.1 NM_026061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900015582.mm.1 TC1900001777.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 636194 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mbph4:80/100.66.79.216:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0