This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194116_m1
See other SGSM1 GE Assays ›
Gene Symbol:
Sgsm1
Gene Name:
small G protein signaling modulator 1
Gene Aliases:
2410098H20Rik, AA408781, D5Bwg1524e, E130107M15, NuIP, Rutbc2
Chromosome Location:
Chr.5: 113243220 - 113310787 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194116_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sgsm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162965.1 NP_001156437.1 3 - 4 232 105
NM_001254731.1 NP_001241660.1 3 - 4 232 105
NM_172718.3 NP_766306.2 3 - 4 232 105
GenBank mRNA AK044716.1 - 3 - 4 186 105
AK053555.1 - 3 - 4 166 105
AK170021.1 - 3 - 4 224 105
BC138049.1 - 3 - 4 158 8861068 105
BC138050.1 - 3 - 4 158 8861069 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003028.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sgsm1

Panther Classification:

Molecular Function -

GTPase-activating protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500026987.mm.1 TC0500003028.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026987.mm.1 NM_001162965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026987.mm.1 NM_001162965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026987.mm.1 NM_001254731.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026987.mm.1 NM_001254731.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026987.mm.1 NM_172718.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026987.mm.1 NM_172718.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500026986.mm.1 TC0500003028.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026986.mm.1 NM_001162965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026986.mm.1 NM_001162965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026986.mm.1 NM_001254731.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026986.mm.1 NM_001254731.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026986.mm.1 NM_172718.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026986.mm.1 NM_172718.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500026988.mm.1 TC0500003028.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026988.mm.1 NM_001162965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026988.mm.1 NM_001162965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026988.mm.1 NM_001254731.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026988.mm.1 NM_001254731.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026988.mm.1 NM_172718.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026988.mm.1 NM_172718.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500026993.mm.1 TC0500003028.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026993.mm.1 NM_001162965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026993.mm.1 NM_001162965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026993.mm.1 NM_001254731.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026993.mm.1 NM_001254731.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026993.mm.1 NM_172718.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026993.mm.1 NM_172718.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9w5rb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0