This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194494_m1
See other EXT1 GE Assays ›
Gene Symbol:
Ext1
Gene Name:
exostoses (multiple) 1
Gene Aliases:
AA409028
Chromosome Location:
Chr.15: 53068261 - 53346183 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194494_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ext1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010162.2 NP_034292.2 9 - 10 2693 78
GenBank mRNA AK132327.1 - 9 - 10 2670 78
BC004741.1 - 9 - 10 2057 3499743 78
U78539.1 - 9 - 10 2197 78
X96639.1 - 9 - 10 2464 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001511.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ext1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

ossification
glycosaminoglycan biosynthetic process
gastrulation
axon guidance
brain development
endoderm development
mesoderm development
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
olfactory bulb development
cellular polysaccharide biosynthetic process
embryonic skeletal joint development
acetylglucosaminyltransferase activity
glucuronosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
protein homodimerization activity
metal ion binding
protein heterodimerization activity
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500011462.mm.1 NM_010162.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011462.mm.1 NM_010162.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011462.mm.1 TC1500001511.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0