This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194544_mH
See other ORAI1 GE Assays ›
Gene Symbol:
Orai1
Gene Name:
ORAI calcium release-activated calcium modulator 1
Gene Aliases:
D730049H07Rik, Tmem142a, orai-1
Chromosome Location:
Chr.5: 123015074 - 123030452 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194544_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Orai1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175423.3 NP_780632.1 1 - 2 489 103
GenBank mRNA AB271214.1 - 1 - 2 305 103
AK052573.1 - 1 - 2 490 103
AK085755.1 - 1 - 2 240 103
AM712355.1 - 1 - 2 305 103
BC023149.1 - 1 - 2 261 4022503 103

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Orai1

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
immune system process
transport
ion transport
calcium ion transport
regulation of calcium ion transport
positive regulation of calcium ion transport
calcium ion transmembrane transport
calcium channel activity
protein binding
calmodulin binding
store-operated calcium channel activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0