This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194715_m1
See other ENPP6 GE Assays ›
Gene Symbol:
Enpp6
Gene Name:
ectonucleotide pyrophosphatase/phosphodiesterase 6
Gene Aliases:
4833421B01Rik, B830047L21Rik, D8Ertd514e, E-NPP 6, Npp6
Chromosome Location:
Chr.8: 46939199 - 47096358 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194715_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Enpp6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177304.3 NP_796278.1 1 - 2 291 81
GenBank mRNA AK038460.1 - 1 - 2 263 81
AK046797.1 - 1 - 2 292 81
AK046881.1 - 1 - 2 296 81
AK079352.1 - 1 - 2 273 81
AK079642.1 - 1 - 2 290 81
BC096376.1 - 1 - 2 262 4237600 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000543.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Enpp6

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
metabolic process
lipid catabolic process
choline metabolic process
catalytic activity
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
photoreceptor cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
glycerophosphodiester phosphodiesterase activity
hydrolase activity
7,8-dihydro-D-neopterin 2',3'-cyclic phosphate phosphodiesterase activity
glycerophosphocholine cholinephosphodiesterase activity
inositol phosphosphingolipid phospholipase activity
inositol phosphorylceramide phospholipase activity
mannosyl-inositol phosphorylceramide phospholipase activity
mannosyl-diinositol phosphorylceramide phospholipase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800003865.mm.1 NM_177304.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800003865.mm.1 TC0800000543.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800003866.mm.1 NM_177304.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800003866.mm.1 TC0800000543.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800003864.mm.1 NM_177304.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800003864.mm.1 TC0800000543.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 714168 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0