This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01195155_g1
See other GPSM2 GE Assays ›
Gene Symbol:
Gpsm2
Gene Name:
G-protein signalling modulator 2 (AGS3-like, C. elegans)
Gene Aliases:
6230410J09Rik, LGN, Pins
Chromosome Location:
Chr.3: 108678638 - 108722330 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01195155_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpsm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029522.2 NP_083798.2 3 - 4 783 70
GenBank mRNA AK040996.1 - 3 - 4 672 70
AK143552.1 - 3 - 4 784 70
AY081187.1 - 3 - 4 627 70
BC021308.1 - 2 - 3 338 5007832 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002750.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpsm2

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
lung epithelial cell differentiation
protein binding
GTPase regulator activity
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300021640.mm.1 NM_029522.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021640.mm.1 NM_029522.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300021640.mm.1 TC0300002750.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300021642.mm.1 NM_029522.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300021642.mm.1 NM_029522.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300021642.mm.1 TC0300002750.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zl4nj:80/100.66.77.81:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0