This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01195248_m1
See other TMPRSS6 GE Assays ›
Gene Symbol:
Tmprss6
Gene Name:
transmembrane serine protease 6
Gene Aliases:
1300008A22Rik
Chromosome Location:
Chr.15: 78439667 - 78468648 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01195248_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027902.2 NP_082178.2 10 - 11 1434 96
GenBank mRNA AK004939.2 - 10 - 11 1281 96
AY240929.1 - 10 - 11 1411 96
BC029645.1 - 10 - 11 1411 4195486 96
BC057674.1 - 10 - 11 1376 4162007 96
EU190436.1 - 9 - 10 1225 96
EU190437.1 - 9 - 10 1225 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001832.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss6

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
negative regulation of BMP signaling pathway
membrane protein proteolysis
fibrinolysis
negative regulation of transcription, DNA-templated
iron ion homeostasis
self proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500014322.mm.1 NM_027902.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014322.mm.1 NM_027902.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500014322.mm.1 TC1500001832.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-8rs9g:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline