This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01195712_g1
See other PLA2G4C GE Assays ›
Gene Symbol:
Pla2g4c
Gene Name:
phospholipase A2, group IVC (cytosolic, calcium-independent)
Gene Aliases:
D7Ertd445e
Chromosome Location:
Chr.7: 13311522 - 13360674 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01195712_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g4c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004762.3 NP_001004762.1 2 - 3 209 60
NM_001168504.1 NP_001161976.1 2 - 3 225 60
GenBank mRNA AK136098.1 - 2 - 3 210 60
AK136143.1 - 2 - 3 212 60
AK145339.1 - 2 - 3 210 60
AK162196.1 - 2 - 3 212 60
AY694793.1 - 2 - 3 196 60
BC117808.1 - 2 - 3 225 40092567 60
BC117809.1 - 2 - 3 225 40092569 60
BC129974.1 - 2 - 3 225 40092571 60
BC138548.1 - 2 - 3 185 8861568 60
BC145767.1 - 2 - 3 185 40131070 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000162.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pla2g4c

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

glycerophospholipid catabolic process
calcium-independent phospholipase A2 activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700001298.mm.1 NM_001004762.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001298.mm.1 NM_001004762.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700001298.mm.1 NM_001168504.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001298.mm.1 NM_001168504.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001298.mm.1 TC0700000162.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700001296.mm.1 NM_001004762.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001296.mm.1 NM_001004762.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700001296.mm.1 TC0700000162.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline