This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01196024_m1
See other IDE GE Assays ›
Gene Symbol:
Ide
Gene Name:
insulin degrading enzyme
Gene Aliases:
1300012G03Rik, 4833415K22Rik, AA675336, AI507533
Chromosome Location:
Chr.19: 37268741 - 37334544 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01196024_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ide
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031156.3 NP_112419.3 21 - 22 2848 142
GenBank mRNA AJ278422.1 - 21 - 22 2868 142
AK004972.2 - 21 - 22 2850 142
AK128986.1 - 21 - 22 2865 142
BC041675.1 - 21 - 22 2880 4951971 142

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ide

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
metabolic process
determination of adult lifespan
bradykinin catabolic process
ubiquitin homeostasis
positive regulation of protein oligomerization
hormone catabolic process
cellular protein catabolic process
negative regulation of proteolysis
beta-amyloid metabolic process
protein homooligomerization
protein homotetramerization
protein heterooligomerization
proteolysis involved in cellular protein catabolic process
insulin metabolic process
insulin catabolic process
nucleotide binding
beta-amyloid binding
glycoprotein binding
catalytic activity
metalloendopeptidase activity
receptor binding
ATP binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
ATPase activity
peptide hormone binding
beta-endorphin binding
peptide binding
protein homodimerization activity
ubiquitin binding
insulin binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline