This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01197658_m1
See other MS4A7 GE Assays ›
Gene Symbol:
Ms4a7
Gene Name:
membrane-spanning 4-domains, subfamily A, member 7
Gene Aliases:
9130422I10Rik, A430103C15Rik, CD20l4, CFFMA
Chromosome Location:
Chr.19: 11321039 - 11336146 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01197658_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ms4a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025610.4 NP_001020781.1 5 - 6 731 151
NM_001276398.1 NP_001263327.1 6 - 7 1028 151
NM_027836.6 NP_082112.3 6 - 7 788 151
GenBank mRNA AF237917.2 - 6 - 7 739 151
AK018686.1 - 5 - 6 702 151
AK079888.1 - 6 - 7 1029 151
AK149890.1 - 6 - 7 756 151
AK151261.1 - 6 - 7 759 151
AK152098.1 - 6 - 7 759 151
BC019784.1 - 5 - 6 677 151
BC024402.1 - 6 - 7 747 5035752 151

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001131.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ms4a7

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958165 BC024402.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958165 AK079888.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958165 AK149890.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958165 AK151261.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958165 BC019784.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135921 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751692 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467996 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394681 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859848 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661286 AK052552.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418684 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12518 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476095 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65254 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104752 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101034 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958165 BC024402.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958165 AK079888.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958165 AK149890.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958165 AK151261.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958165 BC019784.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955593 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955584 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955602 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955588 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955594 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955591 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955608 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955587 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955578 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955599 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955606 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955581 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955597 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955603 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955609 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955586 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955590 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955601 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955585 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955595 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31955580 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104752 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09106136 - -
Whole Mouse Genome G4122A A_51_P277216 BC024402.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P277216 BC019784.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P277216 AF237917.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P59124 AK079888.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P59124 AK149890.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P59124 AK151261.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P59124 AK152098.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P59124 AK018686.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119936 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107046 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104752 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14163579 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14281901 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14163578 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14163588 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14163583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955553 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955586 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955539 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955568 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955580 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955546 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955554 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955493 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955489 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955540 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955549 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955551 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955556 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955591 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955497 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955599 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955523 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955481 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955574 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955527 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955559 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955499 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31955535 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955608 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955581 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955535 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955493 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955611 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955490 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955597 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955570 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955574 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14163579 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955480 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955499 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955491 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955573 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955606 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955486 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955568 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955601 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955599 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955539 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955584 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955587 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955564 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955487 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955497 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955489 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955593 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14346759 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955537 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14316595 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14300257 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955580 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955566 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955590 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955476 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955479 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955560 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955571 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955600 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955482 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955496 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955609 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14346767 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955481 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955585 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14449632 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955559 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955614 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955591 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955484 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14346755 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14497485 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14384366 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955528 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955589 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955472 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955563 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955540 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31955532 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10466207 - -
Affymetrix Mouse Gene 1.0 ST Array 10466208 - -
Affymetrix Mouse Gene 1.0 ST Array 10466209 - -
Affymetrix Mouse Gene 1.0 ST Array 10466205 - -
Affymetrix Mouse Gene 1.0 ST Array 10466204 - -
Affymetrix Mouse Gene 1.0 ST Array 10466201 - -
Affymetrix Mouse Gene 1.0 ST Array 10466202 - -
Affymetrix Mouse Gene 1.0 ST Array 10466203 - -
Affymetrix Mouse Gene 1.0 ST Array 10466206 - -
Affymetrix HT MG-430 PM Array Plate 1424754_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1445365_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10466200 - -
Mouse Genome 430 2.0 Array 1424754_PM_at - -
Mouse Genome 430 2.0 Array 1445365_at - -
Mouse Genome 430 2.0 Array 1424754_at - -
Mouse Genome 430 2.0 Array 1445365_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 109225_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 109225_PM_at - -
Affymetrix Mouse Clariom S Array PSR1900010632.mm.1 NM_001025610.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900010632.mm.1 NM_001276398.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900010632.mm.1 NM_027836.6 Refseq
Affymetrix Mouse Clariom S Array PSR1900010632.mm.1 TC1900001131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900010636.mm.1 NM_001025610.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900010636.mm.1 NM_001276398.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900010636.mm.1 NM_027836.6 Refseq
Affymetrix Mouse Clariom S Array PSR1900010636.mm.1 TC1900001131.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900010637.mm.1 NM_001025610.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900010637.mm.1 NM_001276398.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900010637.mm.1 NM_027836.6 Refseq
Affymetrix Mouse Clariom S Array PSR1900010637.mm.1 TC1900001131.mm.1 MTA TC
Mouse Expression Set 430 1424754_PM_at - -
Mouse Expression Set 430 1424754_at - -
Mouse Expression Set 430 1445365_at - -
Mouse Expression Set 430 1445365_PM_at - -
Murine Genome U74v2 Set 139422_PM_at - -
Murine Genome U74v2 Set 139422_at - -
Mouse Genome 430A 2.0 Array 1424754_at - -
Mouse Genome 430A 2.0 Array 1424754_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1424754_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1424754_at - -
Affymetrix HT Mouse Genome 430B Array 1445365_at - -
Affymetrix HT Mouse Genome 430B Array 1445365_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100380358 - -
Illumina mouse-6 v1.0 expression beadchip 101050280 - -
Illumina mouse-6 v1.0 expression beadchip 2260465 - -
Illumina mouse-6 v1.0 expression beadchip 100380358 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002260465 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2622436 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2622436 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2545828 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1252361 - -
mouse-6 v1.1 (Illumina) scl52727.6_19-S - -
Illumina Sentrix mouse-6.v1 bead array 100380358 - -
Illumina Sentrix mouse-6.v1 bead array 101050280 - -
Illumina Sentrix mouse-6.v1 bead array 2260465 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl52727.6_19-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 101050280 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260465 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 100380358 - -
MouseRef-8 v2 Expression BeadChip ILMN_3091003 - -
MouseRef-8 v2 Expression BeadChip ILMN_3019158 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252361 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3091003 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3019158 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2622436 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0